Assay Details
Gene Symbol: brp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043070.1 | NP_001036535.1 | 3 - 4 | 832 | 77 | |
NM_001103771.4 | NP_001097241.2 | 6 - 7 | 1733 | 77 | ||
NM_001259299.3 | NP_001246228.1 | 6 - 7 | 1733 | 77 | ||
NM_001259300.2 | NP_001246229.1 | 6 - 7 | 1733 | 77 | ||
NM_001259301.2 | NP_001246230.1 | 6 - 7 | 1733 | 77 | ||
NM_001273897.2 | NP_001260826.1 | 5 - 6 | 1342 | 77 | ||
NM_165677.4 | NP_724796.3 | 7 - 8 | 1977 | 77 | ||
NM_206063.3 | NP_995785.2 | 5 - 6 | 1342 | 77 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
