Assay Details
Gene Symbol: Not1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103772.3 | NP_001097242.1 | 1 - 2 | 824 | 86 | |
NM_001103773.3 | NP_001097243.1 | 1 - 2 | 824 | 86 | ||
NM_001169628.2 | NP_001163099.1 | 1 - 2 | 824 | 86 | ||
NM_165678.4 | NP_724798.3 | 1 - 2 | 824 | 86 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
