Assay Details
| Gene Symbol: Urod | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136657.3 | NP_610501.1 | 2 - 3 | 905 | 72 | |
| GenBank mRNA | BT044350.1 | - | 2 - 3 | 881 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Urod| Gene Symbol: Urod | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136657.3 | NP_610501.1 | 2 - 3 | 905 | 72 | |
| GenBank mRNA | BT044350.1 | - | 2 - 3 | 881 | 72 | |
Set Membership: |
|