Assay Details
| Gene Symbol: Sec24AB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299328.1 | NP_001286257.1 | 4 - 5 | 2628 | 66 | |
| NM_001299329.1 | NP_001286258.1 | 5 - 6 | 2259 | 66 | ||
| NM_136687.5 | NP_610531.1 | 4 - 5 | 2325 | 66 | ||
More Information
Set Membership: |
|

Sec24AB
Genomic Map