Assay Details
| Gene Symbol: Sec24AB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299328.1 | NP_001286257.1 | 8 - 9 | 3809 | 88 | |
| NM_001299329.1 | NP_001286258.1 | 9 - 10 | 3440 | 88 | ||
| NM_136687.5 | NP_610531.1 | 8 - 9 | 3506 | 88 | ||
More Information
Set Membership: |
|

Sec24AB
Genomic Map