Assay Details
Gene Symbol: Pal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144158.3 | NP_001137630.2 | 1 - 2 | 226 | 84 | |
NM_001299330.1 | NP_001286259.1 | 1 - 2 | 226 | 84 | ||
NM_136693.3 | NP_610537.2 | 3 - 4 | 612 | 84 | ||
GenBank mRNA | BT050472.1 | - | 3 - 4 | 612 | 84 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
