Assay Details
| Gene Symbol: Pal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144158.3 | NP_001137630.2 | 4 - 5 | 1149 | 65 | |
| NM_001299330.1 | NP_001286259.1 | 4 - 5 | 1149 | 65 | ||
| NM_136693.3 | NP_610537.2 | 6 - 7 | 1535 | 65 | ||
| GenBank mRNA | BT050472.1 | - | 6 - 7 | 1535 | 65 | |
More Information
Set Membership: |
|

Pal1
Genomic Map