Assay Details
| Gene Symbol: Pal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144158.3 | NP_001137630.2 | 5 - 6 | 1576 | 63 | |
| NM_136693.3 | NP_610537.2 | 7 - 8 | 1962 | 63 | ||
| GenBank mRNA | BT050472.1 | - | 7 - 8 | 1962 | 63 | |
More Information
Set Membership: |
|

Pal1
Genomic Map