Assay Details
| Gene Symbol: mlt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273924.1 | NP_001260853.1 | 1 | 2077 | 64 | |
| NM_136718.3 | NP_610562.1 | 1 | 2077 | 64 | ||
| NM_165754.2 | NP_724901.1 | 1 | 2424 | 64 | ||
| GenBank mRNA | AY051458.1 | - | 1 | 2056 | 64 | |
More Information
Set Membership: |
|

mlt
Genomic Map