Assay Details
| Gene Symbol: Obp46a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136730.1 | NP_610574.1 | 2 - 3 | 233 | 68 | |
| GenBank mRNA | BT122055.1 | - | 2 - 3 | 233 | 68 | |
| BT132843.1 | - | 2 - 3 | 233 | 68 | ||
More Information
Set Membership: |
|

Obp46a
Genomic Map