Assay Details
| Gene Symbol: SLO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103788.3 | NP_001097258.2 | 17 - 18 | 3517 | 61 | |
| NM_001103789.3 | NP_001097259.2 | 18 - 19 | 3829 | 61 | ||
| NM_001201971.2 | NP_001188900.1 | 17 - 18 | 3813 | 61 | ||
| NM_001201972.3 | NP_001188901.1 | 17 - 18 | 4030 | 61 | ||
| NM_001273932.1 | NP_001260861.1 | 17 - 18 | 4030 | 61 | ||
| NM_001273933.1 | NP_001260862.1 | 16 - 17 | 3020 | 61 | ||
| NM_001273934.1 | NP_001260863.1 | 17 - 18 | 3509 | 61 | ||
| NM_001273935.1 | NP_001260864.1 | 17 - 18 | 4030 | 61 | ||
More Information
Set Membership: |
|

SLO2
Genomic Map