Assay Details
Gene Symbol: SLO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273934.1 | NP_001260863.1 | 23 - 24 | 4434 | 84 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
