Assay Details
| Gene Symbol: SLO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103789.3 | NP_001097259.2 | 15 - 16 | 2818 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLO2| Gene Symbol: SLO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103789.3 | NP_001097259.2 | 15 - 16 | 2818 | 72 | |
Set Membership: |
|