Assay Details
Gene Symbol: CG9003 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103801.2 | NP_001097271.1 | 4 - 5 | 1117 | 62 | |
NM_001103802.2 | NP_001097272.1 | 4 - 5 | 466 | 62 | ||
NM_001103803.3 | NP_001097273.1 | 5 - 6 | 1000 | 62 | ||
NM_001273971.2 | NP_001260900.1 | 5 - 6 | 983 | 62 | ||
NM_001273972.2 | NP_001260901.1 | 5 - 6 | 983 | 62 | ||
NM_001273973.2 | NP_001260902.1 | 5 - 6 | 1000 | 62 | ||
NM_136845.3 | NP_610689.1 | 4 - 5 | 801 | 62 | ||
GenBank mRNA | BT021402.1 | - | 5 - 6 | 1006 | 62 | |
BT120055.1 | - | 4 - 5 | 431 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
