Assay Details
Gene Symbol: CG9003 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103801.2 | NP_001097271.1 | 8 - 9 | 1989 | 90 | |
NM_001103802.2 | NP_001097272.1 | 8 - 9 | 1338 | 90 | ||
NM_001103803.3 | NP_001097273.1 | 9 - 10 | 1872 | 90 | ||
NM_001273971.2 | NP_001260900.1 | 9 - 10 | 1855 | 90 | ||
NM_001273972.2 | NP_001260901.1 | 9 - 10 | 1855 | 90 | ||
NM_001273973.2 | NP_001260902.1 | 9 - 10 | 1872 | 90 | ||
NM_136845.3 | NP_610689.1 | 8 - 9 | 1673 | 90 | ||
GenBank mRNA | BT021402.1 | - | 9 - 10 | 1878 | 90 | |
BT120055.1 | - | 8 - 9 | 1303 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
