Assay Details
| Gene Symbol: Sln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136862.2 | NP_610706.1 | 3 - 4 | 2203 | 63 | |
| GenBank mRNA | BT016038.1 | - | 3 - 4 | 2201 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sln| Gene Symbol: Sln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136862.2 | NP_610706.1 | 3 - 4 | 2203 | 63 | |
| GenBank mRNA | BT016038.1 | - | 3 - 4 | 2201 | 63 | |
Set Membership: |
|