Assay Details
Gene Symbol: Prp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136891.4 | NP_610735.1 | 2 - 3 | 744 | 64 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
