Assay Details
| Gene Symbol: EndoG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136893.4 | NP_610737.1 | 1 | 978 | 113 | |
| GenBank mRNA | AY094821.1 | - | 1 | 955 | 113 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EndoG| Gene Symbol: EndoG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136893.4 | NP_610737.1 | 1 | 978 | 113 | |
| GenBank mRNA | AY094821.1 | - | 1 | 955 | 113 | |
Set Membership: |
|