Assay Details
| Gene Symbol: ClC-b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136954.4 | NP_610798.1 | 3 - 4 | 617 | 83 | |
| GenBank mRNA | AY071581.1 | - | 3 - 4 | 573 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ClC-b| Gene Symbol: ClC-b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136954.4 | NP_610798.1 | 3 - 4 | 617 | 83 | |
| GenBank mRNA | AY071581.1 | - | 3 - 4 | 573 | 83 | |
Set Membership: |
|