Assay Details
| Gene Symbol: Pde8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169803.2 | NP_001163274.1 | 17 - 18 | 2837 | 74 | |
| NM_001169804.2 | NP_001163275.2 | 13 - 14 | 2997 | 74 | ||
| NM_001169805.2 | NP_001163276.1 | 16 - 17 | 2664 | 74 | ||
| NM_001169807.2 | NP_001163278.2 | 16 - 17 | 2950 | 74 | ||
| NM_137970.5 | NP_611814.3 | 17 - 18 | 2668 | 74 | ||
| NM_166616.3 | NP_726350.1 | 16 - 17 | 2950 | 74 | ||
| NM_166617.2 | NP_726351.1 | 14 - 15 | 2084 | 74 | ||
| NM_166618.3 | NP_726352.1 | 15 - 16 | 2433 | 74 | ||
| NM_166619.2 | NP_726353.1 | 4 - 5 | 957 | 74 | ||
| GenBank mRNA | AY060327.1 | - | 4 - 5 | 957 | 74 | |
| BT021326.1 | - | 17 - 18 | 2664 | 74 | ||
| BT133111.1 | - | 15 - 16 | 2433 | 74 | ||
More Information
Set Membership: |
|

Pde8
Genomic Map