Assay Details
| Gene Symbol: Pde8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169803.2 | NP_001163274.1 | 19 - 20 | 3265 | 81 | |
| NM_001169804.2 | NP_001163275.2 | 15 - 16 | 3425 | 81 | ||
| NM_001169805.2 | NP_001163276.1 | 18 - 19 | 3092 | 81 | ||
| NM_001169807.2 | NP_001163278.2 | 18 - 19 | 3378 | 81 | ||
| NM_137970.5 | NP_611814.3 | 19 - 20 | 3096 | 81 | ||
| NM_166616.3 | NP_726350.1 | 18 - 19 | 3378 | 81 | ||
| NM_166617.2 | NP_726351.1 | 16 - 17 | 2512 | 81 | ||
| NM_166618.3 | NP_726352.1 | 17 - 18 | 2861 | 81 | ||
| NM_166619.2 | NP_726353.1 | 6 - 7 | 1385 | 81 | ||
| GenBank mRNA | AY060327.1 | - | 6 - 7 | 1385 | 81 | |
| BT011336.1 | - | 18 - 19 | 3342 | 81 | ||
| BT021326.1 | - | 19 - 20 | 3092 | 81 | ||
| BT133111.1 | - | 17 - 18 | 2861 | 81 | ||
More Information
Set Membership: |
|

Pde8
Genomic Map