Assay Details
| Gene Symbol: Pde8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169803.2 | NP_001163274.1 | 3 - 4 | 933 | 60 | |
| NM_001169805.2 | NP_001163276.1 | 2 - 3 | 760 | 60 | ||
| NM_001169807.2 | NP_001163278.2 | 2 - 3 | 1046 | 60 | ||
| NM_137970.5 | NP_611814.3 | 3 - 4 | 791 | 60 | ||
| NM_166616.3 | NP_726350.1 | 2 - 3 | 1046 | 60 | ||
| GenBank mRNA | BT011336.1 | - | 2 - 3 | 1010 | 60 | |
| BT021326.1 | - | 3 - 4 | 787 | 60 | ||
More Information
Set Membership: |
|

Pde8
Genomic Map