Assay Details
| Gene Symbol: Pde8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169803.2 | NP_001163274.1 | 10 - 11 | 1604 | 53 | |
| NM_001169804.2 | NP_001163275.2 | 6 - 7 | 1764 | 53 | ||
| NM_001169805.2 | NP_001163276.1 | 9 - 10 | 1431 | 53 | ||
| NM_001169807.2 | NP_001163278.2 | 9 - 10 | 1717 | 53 | ||
| NM_137970.5 | NP_611814.3 | 10 - 11 | 1462 | 53 | ||
| NM_166616.3 | NP_726350.1 | 9 - 10 | 1717 | 53 | ||
| NM_166617.2 | NP_726351.1 | 7 - 8 | 851 | 53 | ||
| NM_166618.3 | NP_726352.1 | 8 - 9 | 1200 | 53 | ||
| GenBank mRNA | BT011336.1 | - | 9 - 10 | 1681 | 53 | |
| BT021326.1 | - | 10 - 11 | 1458 | 53 | ||
| BT133111.1 | - | 8 - 9 | 1200 | 53 | ||
More Information
Set Membership: |
|

Pde8
Genomic Map