Assay Details
Gene Symbol: Not11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299884.1 | NP_001286813.1 | 2 - 3 | 663 | 84 | |
NM_138030.4 | NP_611874.1 | 2 - 3 | 456 | 84 | ||
NM_166652.4 | NP_726406.1 | 2 - 3 | 456 | 84 | ||
GenBank mRNA | BT024271.1 | - | 2 - 3 | 547 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
