Assay Details
Gene Symbol: sei | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299885.1 | NP_001286814.1 | 13 - 14 | 2666 | 122 | |
NM_057365.4 | NP_476713.1 | 12 - 13 | 2185 | 122 | ||
NM_166653.2 | NP_726408.1 | 12 - 13 | 2221 | 122 | ||
GenBank mRNA | AY058350.1 | - | 12 - 13 | 2171 | 122 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
