Assay Details
Gene Symbol: sei | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299885.1 | NP_001286814.1 | 14 - 15 | 2875 | 74 | |
NM_057365.4 | NP_476713.1 | 13 - 14 | 2394 | 74 | ||
NM_166653.2 | NP_726408.1 | 13 - 14 | 2430 | 74 | ||
GenBank mRNA | AY058350.1 | - | 13 - 14 | 2380 | 74 | |
U36925.1 | - | 13 - 14 | 2365 | 74 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
