Assay Details
Gene Symbol: sei | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299885.1 | NP_001286814.1 | 15 - 16 | 2968 | 147 | |
NM_057365.4 | NP_476713.1 | 14 - 15 | 2487 | 147 | ||
NM_166653.2 | NP_726408.1 | 14 - 15 | 2523 | 147 | ||
GenBank mRNA | AY058350.1 | - | 14 - 15 | 2473 | 147 | |
U36925.1 | - | 14 - 15 | 2458 | 147 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
