Assay Details
Gene Symbol: sei | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299885.1 | NP_001286814.1 | 8 - 9 | 2105 | 118 | |
NM_057365.4 | NP_476713.1 | 7 - 8 | 1624 | 118 | ||
NM_166653.2 | NP_726408.1 | 7 - 8 | 1660 | 118 | ||
GenBank mRNA | AY058350.1 | - | 7 - 8 | 1610 | 118 | |
U36925.1 | - | 7 - 8 | 1595 | 118 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
