Assay Details
| Gene Symbol: Mlp60A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144278.3 | NP_001137750.1 | 5 - 6 | 560 | 63 | |
| NM_001299892.1 | NP_001286821.1 | 5 - 6 | 970 | 63 | ||
More Information
Set Membership: |
|

Mlp60A
Genomic Map