Assay Details
Gene Symbol: Mlp60A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144278.3 | NP_001137750.1 | 8 - 9 | 1277 | 76 | |
NM_001299892.1 | NP_001286821.1 | 8 - 9 | 1687 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
