Assay Details
Gene Symbol: bs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144279.3 | NP_001137751.1 | 1 - 2 | 1815 | 75 | |
NM_001299908.1 | NP_001286837.1 | 1 - 2 | 1815 | 75 | ||
NM_166667.3 | NP_726438.1 | 1 - 2 | 974 | 75 | ||
GenBank mRNA | AY095071.1 | - | 1 - 2 | 986 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
