Assay Details
| Gene Symbol: bs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299908.1 | NP_001286837.1 | 2 - 3 | 2061 | 70 | |
| NM_166667.3 | NP_726438.1 | 2 - 3 | 1220 | 70 | ||
| GenBank mRNA | AY095071.1 | - | 2 - 3 | 1232 | 70 | |
More Information
Set Membership: |
|

bs
Genomic Map