Assay Details
Gene Symbol: Mmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202070.1 | NP_001188999.1 | 4 - 5 | 1932 | 113 | |
NM_001202071.1 | NP_001189000.1 | 4 - 5 | 1932 | 113 | ||
NM_001202072.2 | NP_001189001.1 | 4 - 5 | 1837 | 113 | ||
NM_001202073.1 | NP_001189002.1 | 4 - 5 | 1932 | 113 | ||
NM_001202074.2 | NP_001189003.1 | 4 - 5 | 1923 | 113 | ||
NM_001202075.1 | NP_001189004.1 | 4 - 5 | 1932 | 113 | ||
NM_001202076.1 | NP_001189005.1 | 4 - 5 | 1932 | 113 | ||
NM_001259570.2 | NP_001246499.1 | 4 - 5 | 1923 | 113 | ||
NM_079128.4 | NP_523852.3 | 4 - 5 | 1923 | 113 | ||
NM_166683.2 | NP_726473.2 | 4 - 5 | 1923 | 113 | ||
GenBank mRNA | AY118405.1 | - | 4 - 5 | 1919 | 113 | |
BT021303.1 | - | 4 - 5 | 1919 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
