Assay Details
| Gene Symbol: Eps-15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138121.2 | NP_611965.2 | 11 - 12 | 3378 | 60 | |
| NM_166686.1 | NP_726481.1 | 10 - 11 | 2937 | 60 | ||
| NM_166687.1 | NP_726482.1 | 10 - 11 | 3315 | 60 | ||
More Information
Set Membership: |
|

Eps-15
Genomic Map