Assay Details
Gene Symbol: Eps-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138121.2 | NP_611965.2 | 10 - 11 | 2736 | 135 | |
NM_166687.1 | NP_726482.1 | 9 - 10 | 2673 | 135 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
