Assay Details
Gene Symbol: NaCP60E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202078.3 | NP_001189007.2 | 18 - 19 | 6466 | 64 | |
NM_001259578.1 | NP_001246507.1 | 15 - 16 | 5567 | 64 | ||
NM_001259579.3 | NP_001246508.1 | 18 - 19 | 6729 | 64 | ||
NM_001259580.2 | NP_001246509.1 | 19 - 20 | 6481 | 64 | ||
NM_001259581.2 | NP_001246510.1 | 18 - 19 | 6729 | 64 | ||
NM_001274243.2 | NP_001261172.1 | 18 - 19 | 6466 | 64 | ||
NM_001274244.2 | NP_001261173.1 | 17 - 18 | 7671 | 64 | ||
NM_001274246.2 | NP_001261175.1 | 18 - 19 | 6466 | 64 | ||
NM_166696.5 | NP_726495.3 | 18 - 19 | 6729 | 64 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
