Assay Details
Gene Symbol: NaCP60E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202078.3 | NP_001189007.2 | 8 - 9 | 2674 | 99 | |
NM_001259578.1 | NP_001246507.1 | 5 - 6 | 1775 | 99 | ||
NM_001259579.3 | NP_001246508.1 | 8 - 9 | 2937 | 99 | ||
NM_001259580.2 | NP_001246509.1 | 8 - 9 | 2674 | 99 | ||
NM_001259581.2 | NP_001246510.1 | 8 - 9 | 2937 | 99 | ||
NM_001274243.2 | NP_001261172.1 | 8 - 9 | 2674 | 99 | ||
NM_001274244.2 | NP_001261173.1 | 8 - 9 | 2674 | 99 | ||
NM_001274245.2 | NP_001261174.1 | 7 - 8 | 2731 | 99 | ||
NM_001274246.2 | NP_001261175.1 | 8 - 9 | 2674 | 99 | ||
NM_166696.5 | NP_726495.3 | 8 - 9 | 2937 | 99 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
