Assay Details
Gene Symbol: NaCP60E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202078.3 | NP_001189007.2 | 14 - 15 | 3731 | 77 | |
NM_001259578.1 | NP_001246507.1 | 11 - 12 | 2832 | 77 | ||
NM_001259579.3 | NP_001246508.1 | 14 - 15 | 3994 | 77 | ||
NM_001259580.2 | NP_001246509.1 | 15 - 16 | 3746 | 77 | ||
NM_001259581.2 | NP_001246510.1 | 14 - 15 | 3994 | 77 | ||
NM_001274243.2 | NP_001261172.1 | 14 - 15 | 3731 | 77 | ||
NM_001274244.2 | NP_001261173.1 | 13 - 14 | 4936 | 77 | ||
NM_001274245.2 | NP_001261174.1 | 13 - 14 | 3788 | 77 | ||
NM_001274246.2 | NP_001261175.1 | 14 - 15 | 3731 | 77 | ||
NM_166696.5 | NP_726495.3 | 14 - 15 | 3994 | 77 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
