Assay Details
| Gene Symbol: zip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014552.3 | NP_001014552.1 | 10 - 11 | 4581 | 97 | |
| NM_001014553.3 | NP_001014553.1 | 10 - 11 | 4517 | 97 | ||
| NM_001202079.1 | NP_001189008.1 | 10 - 11 | 4544 | 97 | ||
| NM_001202080.2 | NP_001189009.1 | 11 - 12 | 4541 | 97 | ||
| NM_001202081.1 | NP_001189010.1 | 10 - 11 | 4538 | 97 | ||
| NM_001202082.2 | NP_001189011.1 | 11 - 12 | 4541 | 97 | ||
| NM_001259587.2 | NP_001246516.1 | 11 - 12 | 4605 | 97 | ||
| NM_079136.4 | NP_523860.2 | 11 - 12 | 4701 | 97 | ||
| NM_166704.4 | NP_726506.1 | 11 - 12 | 4637 | 97 | ||
More Information
Set Membership: |
|

zip
Genomic Map