Assay Details
Gene Symbol: zip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014552.3 | NP_001014552.1 | 2 - 3 | 388 | 85 | |
NM_001014553.3 | NP_001014553.1 | 2 - 3 | 324 | 85 | ||
NM_001202079.1 | NP_001189008.1 | 2 - 3 | 351 | 85 | ||
NM_001202080.2 | NP_001189009.1 | 2 - 3 | 324 | 85 | ||
NM_001202081.1 | NP_001189010.1 | 2 - 3 | 345 | 85 | ||
NM_001202082.2 | NP_001189011.1 | 2 - 3 | 324 | 85 | ||
NM_001259587.2 | NP_001246516.1 | 2 - 3 | 388 | 85 | ||
NM_079136.4 | NP_523860.2 | 2 - 3 | 388 | 85 | ||
NM_166704.4 | NP_726506.1 | 2 - 3 | 324 | 85 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
