Assay Details
Gene Symbol: zip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014552.3 | NP_001014552.1 | 7 - 8 | 1001 | 73 | |
NM_001014553.3 | NP_001014553.1 | 7 - 8 | 937 | 73 | ||
NM_001202079.1 | NP_001189008.1 | 7 - 8 | 964 | 73 | ||
NM_001202080.2 | NP_001189009.1 | 8 - 9 | 961 | 73 | ||
NM_001202081.1 | NP_001189010.1 | 7 - 8 | 958 | 73 | ||
NM_001202082.2 | NP_001189011.1 | 8 - 9 | 961 | 73 | ||
NM_001259587.2 | NP_001246516.1 | 8 - 9 | 1025 | 73 | ||
NM_079136.4 | NP_523860.2 | 8 - 9 | 1121 | 73 | ||
NM_166704.4 | NP_726506.1 | 8 - 9 | 1057 | 73 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
