Assay Details
| Gene Symbol: RpS3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258475.2 | NP_001245404.1 | 1 - 2 | 370 | 92 | |
| NM_079879.4 | NP_524618.1 | 1 - 2 | 370 | 92 | ||
| NM_166714.4 | NP_726518.1 | 2 - 3 | 314 | 92 | ||
| GenBank mRNA | AF034971.1 | - | 1 - 2 | 399 | 92 | |
More Information
Set Membership: |
|

RpS3A
Genomic Map