Assay Details
Gene Symbol: pan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014685.3 | NP_001014685.1 | 12 - 13 | 1846 | 87 | |
NM_001258477.2 | NP_001245406.1 | 11 - 12 | 1599 | 87 | ||
NM_079880.6 | NP_524619.3 | 12 - 13 | 1726 | 87 | ||
NM_166718.3 | NP_726522.1 | 11 - 12 | 1467 | 87 | ||
NM_166720.3 | NP_726524.1 | 11 - 12 | 1433 | 87 | ||
NM_166723.3 | NP_726527.2 | 12 - 13 | 1858 | 87 | ||
NM_166724.4 | NP_726528.2 | 4 - 5 | 3343 | 87 | ||
GenBank mRNA | BT044547.1 | - | 13 - 14 | 1363 | 87 | |
BT053747.1 | - | 11 - 12 | 1335 | 87 | ||
U88538.1 | - | 11 - 12 | 1392 | 87 | ||
Y09125.1 | - | 1574 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
