Assay Details
Gene Symbol: pan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014685.3 | NP_001014685.1 | 9 - 10 | 1432 | 127 | |
NM_001038709.3 | NP_001033798.1 | 9 - 10 | 1432 | 127 | ||
NM_001258476.2 | NP_001245405.1 | 9 - 10 | 1432 | 127 | ||
NM_001258477.2 | NP_001245406.1 | 8 - 9 | 1173 | 127 | ||
NM_079880.6 | NP_524619.3 | 9 - 10 | 1300 | 127 | ||
NM_166718.3 | NP_726522.1 | 8 - 9 | 1041 | 127 | ||
NM_166720.3 | NP_726524.1 | 8 - 9 | 1007 | 127 | ||
NM_166723.3 | NP_726527.2 | 9 - 10 | 1432 | 127 | ||
GenBank mRNA | BT021431.1 | - | 9 - 10 | 1365 | 127 | |
BT044547.1 | - | 10 - 11 | 937 | 127 | ||
BT053747.1 | - | 8 - 9 | 909 | 127 | ||
U88538.1 | - | 8 - 9 | 966 | 127 | ||
Y09125.1 | - | 1148 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
