Assay Details
| Gene Symbol: Crk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_166744.3 | NP_726550.1 | 2 - 3 | 383 | 75 | |
| GenBank mRNA | AY061094.1 | - | 2 - 3 | 265 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crk| Gene Symbol: Crk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_166744.3 | NP_726550.1 | 2 - 3 | 383 | 75 | |
| GenBank mRNA | AY061094.1 | - | 2 - 3 | 265 | 75 | |
Set Membership: |
|