Assay Details
Gene Symbol: CaMKI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079883.3 | NP_524622.1 | 1 - 2 | 1536 | 90 | |
NM_166759.2 | NP_726569.1 | 2 - 3 | 393 | 90 | ||
NM_166760.2 | NP_726570.1 | 3 - 4 | 734 | 90 | ||
NM_166761.2 | NP_726571.2 | 2 - 3 | 803 | 90 | ||
GenBank mRNA | BT001637.1 | - | 3 - 4 | 344 | 90 | |
Y17917.1 | - | 1477 | 90 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
