Assay Details
Gene Symbol: CaMKI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079883.3 | NP_524622.1 | 5 - 6 | 1930 | 128 | |
NM_166759.2 | NP_726569.1 | 6 - 7 | 787 | 128 | ||
NM_166760.2 | NP_726570.1 | 7 - 8 | 1128 | 128 | ||
NM_166761.2 | NP_726571.2 | 5 - 6 | 1152 | 128 | ||
NM_166764.2 | NP_726574.1 | 4 - 5 | 498 | 128 | ||
GenBank mRNA | BT001637.1 | - | 7 - 8 | 738 | 128 | |
Y17917.1 | - | 1871 | 128 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
