Assay Details
Gene Symbol: Eph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169353.2 | NP_001162824.1 | 6 - 7 | 1264 | 106 | |
NM_079886.5 | NP_524625.3 | 5 - 6 | 1338 | 106 | ||
NM_166774.3 | NP_726588.2 | 5 - 6 | 1338 | 106 | ||
NM_166775.4 | NP_726589.2 | 5 - 6 | 1338 | 106 | ||
NM_166776.4 | NP_726590.2 | 5 - 6 | 1338 | 106 | ||
NM_166777.4 | NP_726591.2 | 6 - 7 | 1264 | 106 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
