Assay Details
Gene Symbol: Eph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169353.2 | NP_001162824.1 | 9 - 10 | 2373 | 110 | |
NM_079886.5 | NP_524625.3 | 8 - 9 | 2447 | 110 | ||
NM_166774.3 | NP_726588.2 | 8 - 9 | 2447 | 110 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
