Assay Details
Gene Symbol: ey | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014693.2 | NP_001014693.1 | 3 - 4 | 968 | 93 | |
NM_001014694.2 | NP_001014694.1 | 2 - 3 | 408 | 93 | ||
NM_079889.3 | NP_524628.2 | 2 - 3 | 788 | 93 | ||
NM_166789.2 | NP_726607.1 | 2 - 3 | 360 | 93 | ||
GenBank mRNA | BT011390.1 | - | 2 - 3 | 360 | 93 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BT011390.1, NM_166789.2 transcripts.
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
